Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST PHENYL-5PW
Supplier:  Tosoh Bioscience LLC 0020023
Encompass
Column, Hydrophobic interaction chromatography; Tosoh; TSKgel; BioAssist Phenyl-5PW; PEEK; Media: polymer; Particle: 10u m Pore Size: 1000angstrom pH stability: 2.0 to 12.0; 7.8mm I.D. x 5cm length
Catalog No. 50-851-624