Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST Q 4.6X50 10UM
Supplier:  Tosoh Bioscience LLC 0019685
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel BioAssist Q; Matrix: polymethacrylate; Material: PEEK; Counter ion: Clsup- Particle size: 10u m; Pore size: 4000angstrom pH: 2.0 -12; 4.6mm I.D. x 5cm length
Catalog No. 50-851-578