Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST S 4.6X50 7UM
Supplier:  Tosoh Bioscience LLC 0019686
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel BioAssist S; Matrix: polymethacrylate; Material: PEEK; Counter ion: Nasup+ Particle size: 7u m; Pore size: 1300angstrom pH: 2.0 -12; 4.6mm I.D. x 5cm length
Catalog No. 50-851-579