Learn More
Tosoh Bioscience LLC TSKGEL BIOASSIST SWXL GUARD
Supplier:  Tosoh Bioscience LLC 0018008
Encompass
Column, Guard; For Tosoh TSKgel BioAssist SWxl; Media: Silica; 6mm I.D. x 4cm Length
Catalog No. 50-851-472