Learn More
Tosoh Bioscience LLC TSKGEL BORONATE-5PW GL 5X50
Supplier:  Tosoh Bioscience LLC 0014449
Encompass
Column, Affinity Chromatography; Tosoh; TSKgel; Boronate-5PW; Made of glass; Media: Polymer; Particle: 10u m; Pore size: 1000angstrom pH stability: 2.0 to 9.0; 5mm I.D. x 5cm length
Catalog No. 50-851-345