Learn More
Tosoh Bioscience LLC TSKGEL BUTYL-NPR 4.6X100
Supplier:  Tosoh Bioscience LLC 0042168
Encompass
Column, Hydrophobic Interaction Chromatography; Tosoh TSKgel Butyl-NPR; Media: Polymer; Particle: 2.5 micrometer; Pore Size: N/A; 4.6mm I.D. x 10cm Length; Stainless Steel
Catalog No. 50-851-710