Learn More
Tosoh Bioscience LLC TSKGEL BUTYL-NPR 4.6X35
Catalog No. 50851379 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Nonporous (NPR) HPLC; Hydrophobic Interaction; Tosoh TSKgel Butyl-NPR; Media: Nonporous polymer; Particle: 2.5 micrometer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-379 Supplier Tosoh Bioscience LLC Supplier No. 0014947
Column, Nonporous (NPR) HPLC; Hydrophobic Interaction; Tosoh TSKgel Butyl-NPR; Media: Nonporous polymer; Particle: 2.5 micrometer; 4.6mm I.D. x 3.5cm Length