Learn More
Tosoh Bioscience LLC TSKGEL CHELATE-5PW 7.5X75
Catalog No. 50851279 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Affinity Chromatography; Tosoh TSKgel Chelate-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 7.5mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-279 Supplier Tosoh Bioscience LLC Supplier No. 0008645
Column, Affinity Chromatography; Tosoh TSKgel Chelate-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 7.5mm I.D. x 7.5cm Length; Stainless Steel