Learn More
Tosoh Bioscience LLC TSKGEL CM-3SW 7.5X75
Supplier:  Tosoh Bioscience LLC 0007162
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel CM-3SW; Matrix: spherical silica; Stainless-steel; Counter ion: Nasup+ Particle: 10u m; Pore size: 250angstrom pH: 2.0 -7.5; 7.5mm I.D. x 7.5cm length
Catalog No. 50-851-169