Learn More
Tosoh Bioscience LLC TSKGEL DEAE-2SW 4.6X250
Supplier:  Tosoh Bioscience LLC 0007168
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel DEAE-2SW; Matrix: hydrophilic silica; Stainless-steel; Counter ion: H2; PO4 sup- Particle: 5u m; Pore size: 125angstrom pH:
Catalog No. 50-851-175