Learn More
Tosoh Bioscience LLC TSKGEL DEAE-5PW (20) 25ML
Supplier:  Tosoh Bioscience LLC 0043381
Encompass
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh TSKgel DEAE-5PW (20); Media: Polymer; Container Size: 25mL
Catalog No. 50-851-744