Learn More
Tosoh Bioscience LLC TSKGEL DEAE-5PW 21.5X150
Supplier:  Tosoh Bioscience LLC 0007574
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel DEAE-5PW; Matrix: hydroxylated methacrylate polymer; Stainless-steel; Counter ion: Clsup- Particle: 13u m; Pore size: 1000angstrom pH: 2.0 -12; 21.5mm I.D. x 15
Catalog No. 50-851-212
Add to Cart
Request a Quote