Learn More
Tosoh Bioscience LLC TSKGEL DEAE-5PW (30) 25ML
Supplier: Tosoh Bioscience LLC 0043281
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh TSKgel DEAE-5PW (30); Media: Polymer; Container Size: 25mL