Learn More
Tosoh Bioscience LLC TSKGEL DEAE-5PW 7.5X75
Supplier:  Tosoh Bioscience LLC 0007164
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel DEAE-5PW; Matrix: hydroxylated methacrylate polymer; Stainless-steel; Counter ion: Clsup- Particle: 10u m; Pore size: 1000angstrom pH: 2.0 -12; 7.5mm I.D. x 7.5
Catalog No. 50-851-171