Learn More
Tosoh Bioscience LLC TSKGEL DEAE-NPR 4.6X35
Supplier:  Tosoh Bioscience LLC 0013075
Encompass
Column, Nonporous (NPR) HPLC; Anion Exchange; Tosoh TSKgel DEAE-NPR; Media: Nonporous polymer; Particle: 2.5 micrometer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-303