Learn More
Tosoh Bioscience LLC TSKGEL DEAE-SW GUARDGEL KIT
Supplier:  Tosoh Bioscience LLC 0007648
Encompass
Kit, Guardgel; For Tosoh TSKgel DEAE-2SW and 3SW columns; Media: Silica
Catalog No. 50-851-220