Learn More
Tosoh Bioscience LLC TSKGEL DNA-STAT
Supplier:  Tosoh Bioscience LLC 0021962
Encompass
The TSKgel DNA-STAT column is packed with 5 �m hydrophilic nonporous resin particles of which the surface consists of an open access network of multi-layered quaternary ammonium anion exchange groups. The innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity, low operating pressure and rapid analysis.
Catalog No. NC0009161