Learn More
Tosoh Bioscience LLC TSKGEL ETHER-5PW 2X75
Supplier:  Tosoh Bioscience LLC 0018760
Encompass
Column, Hydrophobic Interaction Chromatography; Tosoh TSKgel Ether-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 2mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-519