Learn More
Tosoh Bioscience LLC TSKGEL ETHER-5PW (30) 250ML
Supplier:  Tosoh Bioscience LLC 0016050
Encompass
Media, Preparative Bulk Resins; For Hydrophobic Interaction Chromatography; Tosoh TSKgel Ether-5PW (30); Media: Polymer; Container Size: 250mL
Catalog No. 50-851-381