Learn More
Tosoh Bioscience LLC TSKGEL ETHER-5PW 7.5X75
Supplier: Tosoh Bioscience LLC 0008641
Column, Hydrophobic Interaction Chromatography; Tosoh TSKgel Ether-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 7.5mm I.D. x 7.5cm Length; Stainless Steel