Learn More
Tosoh Bioscience LLC TSKGEL ETHER-5PW GL 5X50
Supplier:  Tosoh Bioscience LLC 0014013
Encompass
Column, Hydrophobic Interaction Chromatography; Tosoh TSKgel Ether-5PW Glass; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 5mm I.D. x 5cm Length; Glass
Catalog No. 50-851-321