Learn More
Tosoh Bioscience LLC TSKGEL G-DNA-PW 7.8X300
Supplier:  Tosoh Bioscience LLC 0008032
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G-DNA-PW; Media: Polymer; Particle: 10 micrometer; Pore Size: $>$1000 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - Specialty
Catalog No. 50-851-253