Learn More
Tosoh Bioscience LLC TSKGEL G-OLIGO-PW 7.8X300
Supplier:  Tosoh Bioscience LLC 0008031
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G-Oligo-PW; Media: Polymer; Particle: 7 micrometer; Pore Size: 125 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - Specialty.
Catalog No. 50-851-252