Learn More
Tosoh Bioscience LLC TSKGEL G1000HXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0016131
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel G1000HXL; Media: Polymer; Particle: 5 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-392