Learn More
Tosoh Bioscience LLC TSKGEL G2000PW 7.5X300
Supplier: Tosoh Bioscience LLC 0005761
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G2000PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 125 angstrom; 7.5mm I.D. x 30cm Length; Stainless Steel