Learn More
Tosoh Bioscience LLC TSKGEL G2000SW 21.5X300
Supplier:  Tosoh Bioscience LLC 0006727
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G2000SW; Media: Silica; Particle: 10 micrometer; Pore Size: 125 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-150
Only null left
Add to Cart
Request a Quote