Learn More
Tosoh Bioscience LLC TSKGEL G2000SW 7.5X600
Supplier:  Tosoh Bioscience LLC 0005102
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G2000SW; Media: Silica; Particle: 10 micrometer; Pore Size: 125 angstrom; 7.5mm I.D. x 60cm Length; Stainless Steel
Catalog No. 50-851-092