Learn More
Tosoh Bioscience LLC TSKGEL G2500PW 21.5X600
Supplier:  Tosoh Bioscience LLC 0008030
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G2500PW; Media: Polymer; Particle: 17 micrometer; Pore Size: <200 angstrom; 21.5mm I.D. x 60cm Length; Stainless Steel - Standard Anaytical
Catalog No. 50-851-251
Add to Cart
Request a Quote