Learn More
Tosoh Bioscience LLC TSKGEL G2500PW 7.5X600
Supplier:  Tosoh Bioscience LLC 0008029
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G2500PW; Particle: 10 micrometer; Pore Size: <200 angstrom; Media: Polymer; 7.5mm I.D. x 60cm Length; Stainless Steel - Standard Anaytical
Catalog No. 50-851-250