Learn More
Tosoh Bioscience LLC TSKGEL G2500PWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0008020
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G2500PWxl; Media: Polymer; Particle: 6 micrometer; Pore Size: <200 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-241