Learn More
Tosoh Bioscience LLC TSKGEL G3000PW 21.5X300
Supplier:  Tosoh Bioscience LLC 0016249
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G3000PW; Media: Polymer; 21.5mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-408
Only null left
Add to Cart
Request a Quote