Learn More
Tosoh Bioscience LLC HPLC Column TSKgel G3000PW, 200 Å Polymer, 12 µm, 7.5 x 300 mm
Supplier:  Tosoh Bioscience LLC 0005762
Encompass
TSKgel PW series phases are hydrophilic, rigid, spherical, porous methacrylate beads for aqueous SEC: pH range of 2 to 12, with up to 50% polar organic solvent Wide choice of pore sizes for separation ranges up to 2 x 107 Da for linear polymers Linear SEC column line incorporating proprietary multi-pore technology
Catalog No. 50-851-131