Learn More
Tosoh Bioscience LLC TSKGEL G3000PW 7.5X600
Supplier:  Tosoh Bioscience LLC 0005106
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G3000PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 200 angstrom; 7.5mm I.D. x 60cm Length; Stainless Steel
Catalog No. 50-851-096