Learn More
Tosoh Bioscience LLC TSKGEL G3000PWXL-CP COLUMN
Catalog No. NC0045593 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G3000PWxl-CP; Media: Polymer; Particle: 7 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. NC0045593 Supplier Tosoh Bioscience LLC Supplier No. 0021873
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G3000PWxl-CP; Media: Polymer; Particle: 7 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL