Learn More
Tosoh Bioscience LLC TSKGEL G4000PWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0008022
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G4000PWxl; Media: Polymer; Particle: 10 micrometer; Pore Size: 500 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-243