Learn More
Tosoh Bioscience LLC TSKGEL G4000SW 21.5X300
Supplier:  Tosoh Bioscience LLC 0006729
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G4000SW; Media: Silica; Particle: 13 micrometer; Pore Size: 450 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-152
Add to Cart
Request a Quote