Learn More
Tosoh Bioscience LLC TSKGEL G4000SW 7.5X300
Supplier: Tosoh Bioscience LLC 0005790
Column, Gel Filtration Chromatography; Tosoh TSKgel G4000SW; Media: Silica; Particle: 13 micrometer; Pore Size: 450 angstrom; 7.5mm I.D. x 30cm Length; Stainless Steel