Learn More
Tosoh Bioscience LLC TSKGEL G4000SW GL 8X300
Supplier:  Tosoh Bioscience LLC 0008801
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G4000SW Glass; Particle: 13 micrometer; Pore Size: 450 angstrom; Media: Silica; 8mm I.D. x 30cm Length; Glass
Catalog No. 50-851-285