Learn More
Tosoh Bioscience LLC TSKGEL G4000SWXL 7.8X300
Supplier:  Tosoh Bioscience LLC 0008542
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel G4000SWxl; Media: Silica; Particle: 8 micrometer; Pore Size: 450 angstrom; 7.8mm I.D. x 30cm Length
Catalog No. 50-851-270