Learn More
Tosoh Bioscience LLC TSKGEL G6000PWXL 7.8X300
Catalog No. 50851245 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G6000PWxl; Media: Polymer; Particle: 13 micrometer; Pore Size: $>$1000 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-245 Supplier Tosoh Bioscience LLC Supplier No. 0008024
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel G6000PWxl; Media: Polymer; Particle: 13 micrometer; Pore Size: $>$1000 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL