Learn More
Tosoh Bioscience LLC TSKGEL G7000HHR 7.8X300
Supplier:  Tosoh Bioscience LLC 0017359
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel G7000Hhr; Media: Polymer; Particle: 5 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - Multiple Solvent Capability HHR
Catalog No. 50-851-444