Learn More
Tosoh Bioscience LLC TSKGEL GMHHR-N 7.8X300
Supplier:  Tosoh Bioscience LLC 0018055
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel GMHhr-N; Media: Mixed bed polymer; Particle: 5 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - Multiple Solvent Compatibility HHR
Catalog No. 50-851-473