Learn More
Tosoh Bioscience LLC TSKGEL GMHXL-L 7.8X300
Supplier:  Tosoh Bioscience LLC 0016652
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel GMHxl-L; Media: Mixed bed polymer; Particle: 6 micrometer; 7.8mm I.D. x 30cm Length; Stainless Steel - High Resolution XL
Catalog No. 50-851-421