Learn More
Tosoh Bioscience LLC TSKGEL GMPW 7.5X600
Supplier:  Tosoh Bioscience LLC 0008027
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel GMPW; Media: Mixed bed polymer; Particle: 17 micrometer; 7.5mm I.D. x 60cm Length; Stainless Steel - Standard Anaytical
Catalog No. 50-851-248