Learn More
Tosoh Bioscience LLC TSKGEL GRD HHR-L FOR G1000HHR
Supplier:  Tosoh Bioscience LLC 0017368
Encompass
Column, Guard; For Tosoh TSKgel G1000Hhr-G4000Hhr and GMHhr-L; Media: Polymer; Particle: 5 micrometer; 6mm I.D. x 4cm Length
Catalog No. 50-851-447