Learn More
Tosoh Bioscience LLC TSKgel G2000SWxl-G4000SWxl, and QC-PAK GFC Guard Column for 7.8 mm ID columns
Catalog No. 50851271 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Guard; For 7.8mm I.D. Tosoh TSKgel SWxl columns; Media: Silica; Particle: 7 micrometer; 6mm I.D. x 4cm Length
Catalog No. 50-851-271 Supplier Tosoh Bioscience LLC Supplier No. 0008543
Column, Guard; For 7.8mm I.D. Tosoh TSKgel SWxl columns; Media: Silica; Particle: 7 micrometer; 6mm I.D. x 4cm Length