Learn More
Tosoh Bioscience LLC TSKGEL GUARD COLUMN G2000PW
Supplier:  Tosoh Bioscience LLC 0006763
Encompass
Column, Guard; For 7.5mm I.D. Tosoh TSKgel G1000PW & G2000PW; Media: Polymer; Particle: 12 micrometer; 7.5mm I.D. x 7.5cm Length
Catalog No. 50-851-155