Learn More
Tosoh Bioscience LLC TSKGEL GUARD FOR G-OLIGO-PW
Supplier:  Tosoh Bioscience LLC 0008034
Encompass
Column, Guard; For 7.8mm I.D. Tosoh TSKgel G-Oligo-PW column; Media: Polymer; 6mm I.D. x 4cm Length
Catalog No. 50-851-255