Learn More
Tosoh Bioscience LLC TSKGEL GUARD FOR G2500-GMPW
Supplier:  Tosoh Bioscience LLC 0006762
Encompass
Column, Guard; For 7.5mm I.D. Tosoh TSKgel G2500PW-GMPW; Media: Polymer; Particle: 13 micrometer; 7.5mm I.D. x 7.5cm Length
Catalog No. 50-851-154