Learn More
Tosoh Bioscience LLC TSKGEL GUARD FOR SUPERH-L
Supplier:  Tosoh Bioscience LLC 0018002
Encompass
Column, Guard; For Tosoh TSKgel SuperH1000-4000 columns; Media: Polymer; Particle: 3 micrometer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-470